Learn More
VCAM-1 Monoclonal Antibody (1.G11B1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516430
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD106 antibody, clone 1.G11B1 recognizes human VCAM-1, a ~110 kDa molecule whose ligand is VLA4.
VCAM-1 Monoclonal antibody specifically detects VCAM-1 in Human, Porcine, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
VCAM-1 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.09% sodium azide | |
P19320 | |
VCAM1 | |
human endothelial cells | |
100 μg | |
Primary | |
Human, Porcine, Rhesus Monkey | |
Antibody | |
IgG1 |
Flow Cytometry | |
1.G11B1 | |
FITC | |
VCAM1 | |
adhesion molecule VCAM-1; CD106; CD106 antigen; DKFZp779G2333; INCAM100; INCAM-100; MGC99561; sCD106; soluble CD106; soluble VCAM 1; sVCAM 1; vascular cell adhesion molecule; vascular cell adhesion molecule 1; vascular cell adhesion molecule-1; vascular cell adhesion protein 1; V-CAM 1; Vcam1; Vcam-1; VCAM1B | |
Mouse | |
Protein A | |
RUO | |
396925, 712036, 7412 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |