Learn More
CD105 Monoclonal Antibody (3A9), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517041
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17041 targets CD105 in indirect ELISA, FACS, ICC, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-17041 immunogen is purified recombinant fragment of human CD105 expressed in E. Coli. MA5-17041 detects CD105 which has a predicted molecular weight of approximately 71kDa.
CD105 Monoclonal antibody specifically detects CD105 in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CD105 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P17813 | |
ENG | |
Purified recombinant fragment of human CD105 expressed in E. Coli. | |
RUO | |
2022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3A9 | |
Unconjugated | |
ENG | |
AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |