Learn More
CD104 Monoclonal Antibody (450-9D), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180984
286.68 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.01000kg
Description
The epitope recognized by this antibody is reported to be sensitive to formaldehyde fixation and tissue processing. We recommend the use of acetone fixation for frozen sections. A suggested positive control is human tonsil. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD104 antibody, clone 450-9D recognizes the human beta4 integrin, also known as CD104.
CD104 Monoclonal antibody specifically detects CD104 in Human, Mustelid samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD104 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P16144 | |
ITGB4 | |
Purified alpha6 beta4 integrin from A431 cells. | |
200 μg | |
Primary | |
Human, Mustelid | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
450-9D | |
Unconjugated | |
ITGB4 | |
AA407042; C230078O20; CD antigen CD104; CD104; CD104 antigen; GP150; integrin beta 4; integrin beta-4; integrin subunit beta 4; integrin, beta 4; Itgb4 | |
Mouse | |
Protein G | |
RUO | |
3691 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |