missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD103 Monoclonal Antibody (LF61), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516773
Code : NEW
Additional Details : Weight : 0.01000kg
Description
For FACS analysis, use 5 μL of the suggested working dilution to label 10^6 cells in 100 μL.
CD103 Monoclonal antibody specifically detects CD103 in Human samples. It is validated for Flow Cytometry.Specifications
CD103 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 1% BSA and 0.09% sodium azide | |
P38570 | |
Itgae | |
Hairy cell leukaemia cells | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
LF61 | |
FITC | |
Itgae | |
A530055J10; alpha-E1; alpha-M290; aM290; antigen CD103; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; CD103; DC; DCs; HML-1 antigen; human mucosal lymphocyte antigen 1, alpha polypeptide; HUMINAE; integrin alpha E; integrin alpha E, epithelial-associated; integrin alpha E1, epithelial-associated; integrin alpha M290; Integrin alpha-E; Integrin alpha-E heavy chain; Integrin alpha-E light chain; Integrin alpha-IEL; integrin subunit alpha E; integrin, alpha E; integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide); integrin, alpha E, epithelial-associated; Intregrin alpha E2; Itgae; MGC141996; Mucosal lymphocyte 1 antigen | |
Mouse | |
Protein A | |
RUO | |
3682 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |