missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD10 Monoclonal Antibody (MEM-78), PerCP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110064
256.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD10 Monoclonal antibody specifically detects CD10 in Human samples. It is validated for Flow Cytometry.Specifications
CD10 | |
Monoclonal | |
PerCP | |
Mme | |
6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
4311 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-78 | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P08473 | |
Mme | |
NALM-6 human pre-B cell line | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |