Learn More
CCRL2/CRAM-A/B Mouse anti-Human, Clone: 152254, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB23501
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449375
  • £390.72 / 500µg
Estimated Shipment
17-08-2020
Description
Description
CCRL2/CRAM-A/B Monoclonal antibody MAB23501 Specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
CCRL2/CRAM-A/B
152254
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
NP_001124382
ACKR5, C-C chemokine receptor-like 2, CCR11, CCR6, CCRL2, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRX, CKRXMGC116710, CRAM, CRAMA, CRAM-A, CRAMB, CRAM-B, FLJ55815, HCR, HCRMGC34104, Putative MCP-1 chemokine receptor
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CCRL2/CRAM-A/B. Stains human CCRL2-transfected cells but not irrelevant transfectants. Detects both the CRAM-A and CRAM-B isoforms.
Human
Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CCRL2
NS0 mouse myeloma cell line transfected with human CRAM-A Met1-Val356 (Val180Met) Accession # NP_001124382
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9034
Reconstitute at 0.5 mg/mL in sterile PBS.