Learn More
MCP-2 Recombinant Polyclonal Antibody (3HCLC), Invitrogen™
Rabbit Recombinant Polyclonal Antibody
Brand: Invitrogen 710205
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.
MCP-2 Recombinant Polyclonal antibody specifically detects MCP-2 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Specifications
MCP-2 | |
Polyclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8 | |
Recombinant protein corresponding to amino acids 36–161 of human monocyte chemoattractant protein 2 | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
3HCLC | |
Unconjugated | |
P80075 | |
Rabbit | |
Protein A | |
RUO | |
6355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |