Learn More
CCL25/TECK Rat anti-Mouse, Clone: 89827, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB4811
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13430726
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
CCL25/TECK Monoclonal antibody MAB4811 Specifically detects CCL25/TECK in Mouse samples. It is validated for ELISA Capture, Immunohistochemistry, Western Blotting
Specifications
Specifications
CCL25/TECK
89827
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O35903.1
C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse CCL25/TECK in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, and recombinant rat CCL20 is observed.
Mouse
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CCL25
E. coli-derived recombinant mouse CCL25/TECK Gln24-Asn144 Accession # O35903.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
6370
Reconstitute at 0.5 mg/mL in sterile PBS.