Learn More
CCI Mouse anti-Viral, Clone: 112811, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB696
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13461606
  • £337.93 / 500µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
CCI Monoclonal antibody MAB696 Specifically detects CCI in Viral samples. It is validated for Western Blotting
Specifications
Specifications
CCI
112811
Western Blot 1 ug/mL
Purified
S. frugiperda insect ovarian cell line Sf 21-derived recombinant viral CCI
Protein A or G purified from ascites
RUO
Primary
3707616
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects viral CCI in direct ELISAs and Western blots.
Virus