Learn More
Caveolin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA137260
Code : LD
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is lung carcinoma.
Caveolin 1 Polyclonal antibody specifically detects Caveolin 1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Caveolin 1 | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q03135 | |
CAV1 | |
Synthetic peptide from N-terminus of human caveolin-1. | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CAV1 | |
BSCL3; CAV; CAV1; Cav-1; caveolae; caveolin 1; caveolin 1, caveolae protein; caveolin 1, caveolae protein, 22kDa; caveolin 1a; caveolin 1b; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; caveolin-1; caveolin-1a; caveolin-1b; cell growth-inhibiting protein 32; CGL3; HGNC:1527; LCCNS; MSTP085; OTTHUMP00000196475; PPH3; Unknown (protein for IMAGE:7985573); variant a; variant b; vesicular integral-membrane protein VIP21; VIP21; wu:fc07c04 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
857 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |