Learn More
CATIP Monoclonal Antibody (2C3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523285
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MSSKVYSTGS RAKDHQPSGP ECLPLPEANA EAIDFLSSLH KEELQMLFFS ETLAMVSDTG EPQGELTIEV QRGKYQEKLG MLTYCLFVHA SSRGFLDKML CGNSLLGYLS EKLELMEQHS QDFIKFLILP MERKMSLLKQ DDQLAVTRSI KEGEEVKTGV TSFPWSSIKG FISEAANLVL LRVMAWRRMV PSNARFLTLD TEGKLCYLTY QNLGFQTIQV DHQQAEVFIV EQTVHAEEGI PMSCQYYLLS DGHLAKRIQV GSPGCCIITK MPILREEDEI EPRPVFEKKP LVWEEDMELY SKFLDRKEEL RLGHASYLRQ HPEAHALISD FLLFLLLRQP EDVVTFAAEF FGPFDPWRPS SPALGSSHRP NPFRSLEPEG DARSGAA
CATIP Monoclonal antibody specifically detects CATIP in Human samples. It is validated for ELISA.Specifications
CATIP | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q7Z7H3 | |
CATIP | |
CATIP (AAH52750, 1 a.a. ∽ 387 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
2C3 | |
Unconjugated | |
CATIP | |
C15H2orf62; C2orf62; C37H2orf62; C6H2orf62; Catip; ciliogenesis associated TTC17 interacting protein; Ciliogenesis-associated TTC17-interacting protein; Gm216; hypothetical protein LOC690437; LOC690437 | |
Mouse | |
Affinity chromatography | |
RUO | |
375307 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |