Learn More
Cathepsin X/Z/P Mouse anti-Human, Clone: 163702, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB934
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13442016
  • £379.26 / 500µg
Estimated Shipment date
from Supplier 25-10-2019
Description
Description
Cathepsin X/Z/P Monoclonal antibody MAB934 Specifically detects Cathepsin X/Z/P in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Cathepsin X/Z/P
Unconjugated
Murine
IgG2a
500μg
Primary
Human
163702
Purified
Mouse myeloma cell line NS0-derived recombinant human Cathepsin X/Z/P. Ala21-Val303 (Gly23Val, Ser48Thr)
Protein A or G purified from hybridoma culture supernatant