missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin V Mouse anti-Human, Clone: 182021, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB10801

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468315

  • £311.64 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cathepsin V Monoclonal antibody MAB10801 Specifically detects Cathepsin V in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin V
182021
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CTSL2
Mouse myeloma cell line NS0-derived recombinant human Cathepsin V Val18-Val334 Accession # O60911
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1515
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
O60911
cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSL2, CTSUcathepsin L2, preproprotein, CTSVMGC125957, EC 3.4.22, EC 3.4.22.43
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cathepsin V in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Cathepsin B, C, F, K, L, O, S, or Z is observed. In Western blots, less than 5% cross-reactivity with rhCathepsin K and no corss-reactivity with rhCathepsin B, C, F, L, O, S, or Z is observed. This antibody specifically identifies the pro form but not the active form of Cathepsin V in Western blots.
Human
Videos
Documents

For Research Use Only