Learn More
Cathepsin S Rat anti-Human, Clone: 248718, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1183
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448425
  • £390.72 / 500µg
Estimated Shipment
17-08-2020
Description
Description
Cathepsin S Monoclonal antibody MAB1183 Specifically detects Cathepsin S in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Cathepsin S
248718
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CTSS
Mouse myeloma cell line NS0-derived recombinant human Cathepsin S Gln17-Ile331 (predicted) Accession # P25774
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1520
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P25774
cathepsin S, CTSS, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects pro and mature forms of human Cathepsin S in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Cathepsin B, C, D, L, L2/V, X/Z/P, recombinant mouse Cathepsin F, or H is observed.