missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin L Monoclonal Antibody (33/2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA126774
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: mink lung cell.
Cathepsin L Monoclonal antibody specifically detects Cathepsin L in Human, Mink, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Western Blot.Specifications
Cathepsin L | |
Monoclonal | |
4.3 mg/mL | |
Ascites with 15mM sodium azide | |
P06797, P07154, P07711 | |
CTSL | |
Purified, full length, native protein from human lung cancer line EPLC 32 M1. | |
RUO | |
13039, 1514, 25697 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
33/2 | |
Unconjugated | |
CTSL | |
1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L | |
Mouse | |
100 μL | |
Primary | |
Human, Mink, Mouse, Rat | |
Antibody | |
IgG1 |