missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin L Mouse anti-Insect, Clone: 193702, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB22591

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449305

  • £311.64 / 100µg

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cathepsin L Monoclonal antibody MAB22591 Specifically detects Cathepsin L in Insect samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin L
193702
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
cathepsin L, cathepsin L1, CATL, CTSL, CTSL1, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects pro and mature forms of Insect Cathepsin L in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human Cathepsin L is observed and no cross-reactivity with recombinant mouse Cathepsin L is observed.
Insect
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Purified
CTSL1
S. frugiperda ovarian cell line Sf 21-derived Insect Cathepsin L
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1514
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only