missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin H Mouse anti-Human, Rat, Clone: AF14D7, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517253
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control for this product is K562 cells.
This protein is important for the overall degradation of proteins in lysosomes.Specifications
Cathepsin H | |
Monoclonal | |
Unconjugated | |
CTSH | |
Cathepsin H; Cathepsin H mini chain; Cathepsin H heavy chain; Cathepsin H light chain; CPSB; CTSH | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
1512, 25425 | |
-20°C | |
Liquid |
Western Blot | |
AF14D7 | |
HEPES with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide | |
P09668, P00786 | |
CTSH | |
Recombit human protein purified from E.coli (His-Cathepsin H) | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |