missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin E Mouse anti-Human, Clone: 212211, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1294

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438525

  • £337.93 / 500µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cathepsin E Monoclonal antibody MAB1294 Specifically detects Cathepsin E in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin E
212211
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CTSE
Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Gln18-Pro396 Accession # NP_001901
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1510
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_001901
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cathepsin E in direct ELISAs and Western blots. It recognizes both the pro and mature forms of recombinant human (rh) Cathepsin E. In Western blots, 100% cross-reactivity with recombinant mouse (rm) Cathepsin E and recombinant human (rh) BACE is observed under reducing conditions. Under non-reducing conditions, less than 1% cross-reactivity with rmCathepsin E and rhBACE-1 is observed. Less than 1% cross-reactivity with rhCathepsin D is observed under both reducing and non-reducing conditions.
Human
Videos

For Research Use Only