Learn More
Cathepsin E Rat anti-Mouse, Clone: 186513, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1130
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488375
  • £358.00 / 500µg
Estimated Shipment
08-03-2018
Add to basket
Description
Description
Cathepsin E Monoclonal antibody MAB1130 Specifically detects Cathepsin E in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Cathepsin E
Unconjugated
Rat
IgG2a
500μg
Primary
Murine
186513
Purified
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E. Gln19-Pro397
Protein A or G purified from hybridoma culture supernatant