Learn More
Cathepsin D Rat anti-Mouse, Clone: 204712, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1029
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458265
  • £337.93 / 500µg
Estimated Shipment
06-03-2019
Add to basket
Description
Description
Cathepsin D Monoclonal antibody MAB1029 Specifically detects Cathepsin D in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Cathepsin D
204712
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CTSD
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin D Ile21-Leu410 Accession # P18242
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1509
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P18242
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects both the pro and active forms of mouse Cathepsin D in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Cathepsin D or recombinant mouse Cathepsin E is observed.