missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin C Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537849
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Cathepsin C Polyclonal antibody specifically detects Cathepsin C in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Cathepsin C | |
Polyclonal | |
Unconjugated | |
CTSC | |
AI047818; CATC; Cathepsin C; cathepsin J; CPPI; CTSC; Dipeptidyl peptidase 1; Dipeptidyl peptidase 1 exclusion domain chain; Dipeptidyl peptidase 1 heavy chain; Dipeptidyl peptidase 1 light chain; Dipeptidyl peptidase I; Dipeptidyl peptidase I exclusion domain chain; Dipeptidyl peptidase I heavy chain; Dipeptidyl peptidase I light chain; Dipeptidyl transferase; dipeptidyl-peptidase; dipeptidylpeptidase 1; dipeptidyl-peptidase 1; dipeptidyl-peptidase I; DPP1; DPPI; DPP-I; HMS; JP; JPD; PALS; PDON1; PLS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1075 | |
-20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P53634 | |
CTSC | |
Peptide with sequence C-RNVHGINFVSPVRNQ, from the internal region of CTSC (aa238-252) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |