Learn More
Cathepsin A/Lysosomal Carboxypeptidase A Rat anti-Mouse, Clone: 191424, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB11231
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488365
  • £358.00 / 500µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal antibody MAB11231 Specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation
Specifications
Specifications
Cathepsin A/Lysosomal Carboxypeptidase A
Unconjugated
Rat
IgG2b
500μg
Primary
Murine
191424
Purified
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin A. aa 24-474
Protein A or G purified from hybridoma culture supernatant