missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin A/Lysosomal Carboxypeptidase A Mouse anti-Human, Clone: 179803, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1049

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438285

  • £376.32 / 500µg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal antibody MAB1049 Specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin A/Lysosomal Carboxypeptidase A
179803
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CTSA
Mouse myeloma cell line NS0-derived recombinant human Cathepsin A/Lysosomal Carboxypeptidase A Ala29-Tyr480 (predicted) Accession # P10619
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5476
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P10619
beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase
Mouse
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Cathepsin A/Lysosomal Carboxypeptidase A in direct ELISAs and Western blots. In Western blots, detects the single chain (55 kDa) and heavy chain (32 kDa) forms of recombinant human (rh) Cathepsin A. In Western blots, less than 5% cross-reactivity with rhCathepsin B, C, D, E, L, O, S, X and Z is observed and no cross-reactivity with the light chain (20 kDa) of rhCathepsin A is observed.
Human
Videos
Documents

For Research Use Only