Learn More
Cathepsin 7/Cathepsin 1 Rat anti-Mouse, Clone: 228905, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1499
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13428685
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
Cathepsin 7/Cathepsin 1 Monoclonal antibody MAB1499 Specifically detects Cathepsin 7/Cathepsin 1 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Cathepsin 7/Cathepsin 1
228905
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Q91ZF2
Cts7
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 7/Cathepsin 1 Ala18-Leu331 Accession # Q91ZF2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects mouse Cathepsin 7/Cathepsin 1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Cathepsin O, S, or V, or recombinant mouse Cathepsin B, C, H, L, or Z is observed.