Learn More
Cathepsin 6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547263
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse (rm) Cathepsin 3 is observed and 10% cross-reactivity with rmCathepsin 1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cathepsin 6 Polyclonal antibody specifically detects Cathepsin 6 in Mouse samples. It is validated for Immunoprecipitation, Western Blot.Specifications
Cathepsin 6 | |
Polyclonal | |
Unconjugated | |
Cts6 | |
1600022N02Rik; cathepsin 6; Cts6 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
58518 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
0 | |
Cts6 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 6 Leu21-Val334 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |