missing translation for 'onlineSavingsMsg'
Learn More

Cathepsin 3 Rat anti-Mouse, Clone: 264013, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2638

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459595

  • £390.72 / 500µg

Estimated Shipment
18-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cathepsin 3 Monoclonal antibody MAB2638 Specifically detects Cathepsin 3 in Mouse samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Cathepsin 3
264013
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
NP_081182
1600000I23Rik
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Cathepsin 3 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Cathepsin 1, 2, or 6 is observed.
Mouse
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Cts3
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin 3 Ser18-Val332 Accession # NP_081182
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
117066
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only