Learn More
delta Catenin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA139454
Code : LD
Additional Details : Weight : 0.01000kg
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast lobular carcinoma.
delta Catenin Polyclonal antibody specifically detects delta Catenin in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
delta Catenin | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
O60716, P30999 | |
CTNND1 | |
Synthetic peptide corresponding to C-terminus of human p120. | |
1 mL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CTNND1 | |
AA409437; AU019353; cadherin-associated Src substrate; Cadherin-associated Src substrate (CAS); CAS; Catenin (cadherin associated protein) delta 1; catenin (cadherin associated protein), delta 1; catenin (cadherin-associated protein), delta 1; catenin delta 1; catenin delta-1; catenin src; catenin, delta 1; Catns; CTNND; Ctnnd1; hypothetical protein; Kiaa0384; mKIAA0384; p120; p120 catenin; P120 CTN; P120 p120; P120 P120CTN; p120(CAS); p120(ctn); P120CAS; p120-catenin; P120CTN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12388, 1500 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |