missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cathepsin B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514255
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate based on sequence homology.
Cathepsin B Polyclonal antibody specifically detects Cathepsin B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Cathepsin B | |
Polyclonal | |
Unconjugated | |
Ctsb | |
amyloid precursor protein secretase; APP secretase; APPS; cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; CB; CPSB; Ctsb; cysteine protease; RSG-2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1508 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P07858 | |
Ctsb | |
KLH conjugated synthetic peptide between 198-227 amino acids from the central region of human CATB | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |