missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Mouse anti-Human, Mouse, Rat, Clone: 1A1, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0003
Additional Details : Weight : 0.99000kg
Description
A suggested positive control for this product is HeLa cells.
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara′s disease.Specifications
Catalase | |
Monoclonal | |
Unconjugated | |
P04040, P24270, P04762 | |
CAT | |
Purified, recombit, human catalase protein expressed in E.coli. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
ELISA, Immunoprecipitation | |
1A1 | |
CAT | |
CTA1 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
12359, 24248, 847 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |