missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518531
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine based on sequence homology.
Catalase Polyclonal antibody specifically detects Catalase in Human, Mouse samples. It is validated for Western Blot.Specifications
Catalase | |
Polyclonal | |
Unconjugated | |
Cat | |
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
12359, 847 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P04040, P24270 | |
Cat | |
Synthetic peptide sequence (DKYNAEKPKNAIHT) corresponding to the internal amino acids of CAT | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |