missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA129159
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat Cells, Stomach.
Caspase 8 Polyclonal antibody specifically detects Caspase 8 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Caspase 8 | |
Polyclonal | |
Unconjugated | |
Casp8 | |
ALPS2B; apoptosis-related cysteine peptidase; apoptotic cysteine protease; Apoptotic protease Mch-5; CAP4; CASP8; CASP-8; caspase 8; caspase 8, apoptosis-related cysteine peptidase; caspase 8, apoptosis-related cysteine protease; Caspase8; caspase-8; Caspase-8 precursor; Caspase-8 subunit p10; Caspase-8 subunit p18; EC 3.4.22.61; FADD-homologous ICE/ced-3-like protease; FADD-like ICE; Fas-linked ICE-like protease; FLICE; FLJ17672; ICE8; ICE-like apoptotic protease 5; MACH; MACH-alpha-1/2/3 protein; MACH-beta-1/2/3/4 protein; MCH5; MGC78473; MORT1-associated ced-3 homolog; OTTHUMP00000163720; OTTHUMP00000165063; OTTHUMP00000206557; OTTHUMP00000206581 | |
Rabbit | |
Protein A | |
RUO | |
12370, 64044, 841 | |
4° C, do not freeze | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
10mM PBS with 0.2% BSA and 0.1% sodium azide | |
O89110, Q14790, Q9JHX4 | |
Casp8 | |
The epitope corresponding to amino acids 217-350 mapping within the caspase-8 p18 subunit of human origin | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |