missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520110
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is Ramos cell lysate. PA5-20110 can be used with blocking peptide PEP-0228.
Caspase 5 Polyclonal antibody specifically detects Caspase 5 in Human samples. It is validated for Western Blot.Specifications
Caspase 5 | |
Polyclonal | |
Unconjugated | |
CASP5 | |
CASP5; CASP-5; caspase 5; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE(rel)III; ICE(rel)-III; ICEREL-III; ICH3; ICH-3; Protease ICH-3; protease TY; TY protease | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
838 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P51878 | |
CASP5 | |
A 16 amino acid peptide from the amino-terminus of human Caspase-5. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |