missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520109
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human spleen tissue lysate. PA5-20109 can be used with blocking peptide PEP-0227.
Caspase 4 Polyclonal antibody specifically detects Caspase 4 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Caspase 4 | |
Polyclonal | |
Unconjugated | |
CASP4 | |
apoptotic cysteine protease Mih1/TX; Casp11; CASP-11; CASP4; CASP-4; caspase 11; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; Caspase-11; Caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-4 subunit p10; Caspase-4 subunit p20; Caspl; ICE and Ced-3 homolog 2; ICE(rel)II; ICE(rel)-II; ICEREL-II; ICH2; ICH-2; Ich3; ich-3; Mih1; Mih1/TX; protease ICH-2; Protease ICH-3; Protease TX; TX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12363, 837 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P49662, P70343 | |
CASP4 | |
A 15 amino acid peptide from near the center of human Caspase-4. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |