missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 4/5 p20 (Cleaved Asp270, Asp311) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539873
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Caspase 4/5 p20 (Cleaved Asp270, Asp311) Polyclonal antibody specifically detects Caspase 4/5 p20 (Cleaved Asp270, Asp311) in Human samples. It is validated for Western Blot.Specifications
Caspase 4/5 p20 (Cleaved Asp270, Asp311) | |
Polyclonal | |
Unconjugated | |
CASP4 | |
apoptotic cysteine protease Mih1/TX; CASP4; CASP-4; CASP5; CASP-5; caspase 4; caspase 4, apoptosis-related cysteine peptidase; caspase 4, apoptosis-related cysteine protease; caspase 5; caspase 5, apoptosis-related cysteine peptidase; caspase 5, apoptosis-related cysteine protease; caspase-4; Caspase-4 subunit 1; Caspase-4 subunit 2; Caspase-5; Caspase-5 subunit p10; Caspase-5 subunit p20; ICE and Ced-3 homolog 2; ICE(rel)II; ICE(rel)-II; ICE(rel)III; ICE(rel)-III; ICEREL-II; ICEREL-III; ICH2; ICH-2; ICH3; ICH-3; Mih1; Mih1/TX; protease ICH-2; protease ICH-3; protease protease ICE(rel)III; protease protease TY; Protease TX; protease TY; TX; TY protease | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
837, 838 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P49662, P51878 | |
CASP4, CASP5 | |
A synthetic peptide derived from human Caspase 4/5 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |