missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130417
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HL-60 and Jurkat.
Caspase 2 Polyclonal antibody specifically detects Caspase 2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Caspase 2 | |
Polyclonal | |
Unconjugated | |
CASP2 | |
CASP2; CASP-2; caspase 2; caspase 2 apoptosis-related cysteine peptidase; caspase-2; Caspase-2 subunit p12; Caspase-2 subunit p13; Caspase-2 subunit p18; ICH1; ICH-1; ICH-1 protease; ICH-1L; ICH-1L/1S; NEDD2; NEDD-2; Neural precursor cell expressed developmentally down-regulated protein 2; PPP1R57; Protease ICH-1; protein phosphatase 1, regulatory subunit 57 | |
Rabbit | |
Affinity chromatography | |
RUO | |
12366, 64314, 835 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 0.2% gelatin and 0.1% sodium azide | |
P29594, P42575, P55215 | |
CASP2 | |
Synthetic peptide (Human) (C terminal). | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |