missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA520033
Code : LD
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human heart tissue lysate. PA5-20033 can be used with blocking peptide PEP-0152.
Caspase 12 Polyclonal antibody specifically detects Caspase 12 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Caspase 12 | |
Polyclonal | |
Unconjugated | |
CASP12 | |
CASP12; CASP-12; CASP12P1; caspase 12; caspase 12 (gene/pseudogene); caspase 12 pseudogene 1; caspase-12; Inactive caspase-12; OTTMUSP00000045799; OTTMUSP00000045802; OTTMUSP00000045803; UNQ9415; UNQ9415/PRO34398 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100506742, 12364, 156117 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O08736, Q6UXS9, Q920D5 | |
CASP12 | |
A synthetic peptide corresponding to amino acids within the large cleavage product of murine caspase-12. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |