missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CK2 beta Mouse anti-Human, Mouse, Rat, Clone: 26C3, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0216
Additional Details : Weight : 0.99000kg
Description
A suggested positive control for this product is K562 cells.
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.Specifications
CK2 beta | |
Monoclonal | |
Unconjugated | |
P67870, P67871, P67874 | |
CSNK2B | |
Recombit human protein purified from E.coli (His-casein kinase II beta) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 λ |
ELISA, Western Blot | |
26C3 | |
CSNK2B | |
DADB-127H9.2, CK2B, CK2N, CSK2B, G5A, MGC138222, MGC138224 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
13001, 1460, 81650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |