Learn More
CK2 beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527416
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: MCF-7, NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Pig (100%), Rabbit (100%), Chicken (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CK2 beta Polyclonal antibody specifically detects CK2 beta in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CK2 beta | |
Polyclonal | |
Unconjugated | |
Csnk2b | |
alternative name: G5a, phosvitin; casein kinase 2 beta; casein kinase 2, beta polypeptide; casein kinase 2, beta subunit; casein kinase II b subunit splicing isoform 132; casein kinase II b subunit splicing isoform 247; casein kinase II b subunit splicing isoform 318; casein kinase II b subunit splicing isoform 660; casein kinase II b subunit splicing isoform 806; Casein kinase II beta subunit; casein kinase II subunit beta; chimera CSNK2B-LY6G5B splicing isoform 1072; chimera CSNK2B-LY6G5B splicing isoform 1103; chimera Csnk2b-Ly6g5b splicing isoform 2050; chimera Csnk2b-Ly6g5b splicing isoform 2275; chimera Csnk2b-Ly6g5b splicing isoform 2531; chimera CSNK2B-LY6G5B splicing isoform 532; chimera CSNK2B-LY6G5B splicing isoform 560; chimera CSNK2B-LY6G5B splicing isoform 562; chimera Csnk2b-Ly6g5b splicing isoform 901; CK II beta; Ck2; CK2B; CK2N; CSK2B; Csnk2b; Csnk2b splicing isoform 280; Csnk2b splicing isoform 662; Csnk2b-Ly6g5b; CSNK2B-LY6G5B-1072; CSNK2B-LY6G5B-1103; CSNK2B-LY6G5B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13001, 1460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P67870, P67871 | |
Csnk2b | |
Recombinant fragment corresponding to a region within amino acids 1 and 215 of Human Casein Kinase 2 beta | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |