missing translation for 'onlineSavingsMsg'
Learn More

CARS2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA514999

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 10649740

  • £332.00 / 100µg

Estimated Shipment date
from Supplier 09-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CARS is a class 2 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. It catalyzes the chemical reaction: ATP + L-cysteine + tRNA(Cys) = AMP + diphosphate + L-cysteinyl-tRNA(Cys).
Specifications

Specifications

CARS2
Polyclonal
PBS with 0.09% sodium azide
Q9HA77
Probable cysteinyl-tRNA synthetase, mitochondrial; Cysteine--tRNA ligase; CysRS; CARS2; OK/SW-cl.10
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79587
Immunohistochemistry, Western Blot
Unconjugated
CARS2
Liquid
CARS2
KLH conjugated synthetic peptide between 536-564 amino acids from the C-terminal region of human CARS2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Videos
Documents