Learn More
Caronte Mouse anti-Chicken, Clone: 156808, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1051
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448285
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 24-02-2020
Description
Description
Caronte Monoclonal antibody MAB1051 Specifically detects Caronte in Chicken samples. It is validated for Western Blotting
Specifications
Specifications
Caronte
156808
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects chicken Caronte in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Cerberus, rhDAN, or recombinant mouse Gremlin is observed.
Chicken
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_990154
CER1
Mouse myeloma cell line NS0-derived recombinant chicken Caronte Asp16-Asn272 Accession # NP_990154
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9350
Reconstitute at 0.5 mg/mL in sterile PBS.