missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cardiac Troponin I Monoclonal Antibody (415), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191566
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-91566 detects Cardiac Troponin I from human samples.
Cardiac Troponin I Monoclonal antibody specifically detects Cardiac Troponin I in Human samples. It is validated for Affinity Purification, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
Cardiac Troponin I | |
Monoclonal | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P19429 | |
TNNI3 | |
Free human cardiac troponin I, native human cardiac troponin complex, or human cardiac troponin I peptides | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Affinity Purification, Immunohistochemistry, Immunoprecipitation, Western Blot | |
415 | |
Unconjugated | |
TNNI3 | |
alternatively spliced; cardiac ankyrin repeat kinase; Cardiac muscle troponin T; cardiac troponin I; cardiac troponin I isoform; cardiac troponin I-interacting kinase; cardiac troponin T; cardiac troponin T isoform 1; cardiac troponin T isoform 2; cardiac troponin T2; cardiomyopathy, dilated 1D (autosomal dominant); cardiomyopathy, dilated 2A (autosomal recessive); cardiomyopathy, hypertrophic 2; CARK; CCDD; CMD1D; CMD1FF; CMD2A; CMH2; CMH7; CMPD2; cTnI; cTnT; Ctt; CTTG; D830019J24Rik; LVNC6; MGC116817; RATCTTG; RCM1; RCM3; Serine/threonine-protein kinase TNNI3K; Tn1; TnI; TNNC1; Tnni3; TNNI3 interacting kinase; TNNI3-interacting kinase; TNNI3K; tnni3k {ECO:0000250; TNNT2; Tnnt3; TnTc; Tpn1; troponin 1, type 3; troponin I type 3 (cardiac); troponin I, cardiac 3; troponin I, cardiac muscle; troponin I, cardiac muscle-like; troponin I3, cardiac type; Troponin T cardiac; troponin T cardiac isoform; troponin T isoform 1; troponin T isoform 2; troponin T splicing mutant isoform 1; troponin | |
Mouse | |
Protein A | |
RUO | |
7137 | |
4° C, do not freeze | |
Liquid |