missing translation for 'onlineSavingsMsg'
Learn More

CEA Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA137416

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11501753

  • £288.00 / 1mL

Estimated Shipment date
from Supplier 25-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A suggested positive control is colon carcinoma.
Specifications

Specifications

CEA
Polyclonal
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P06731
CD66e, CEA, meconium antigen 100
Rabbit
IgG
1 mL
4° C, do not freeze
Primary
1048
Immunohistochemistry (Paraffin)
Unconjugated
CEACAM5
Liquid
CEACAM5
Purified human CEA.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documents