Learn More
Carboxypeptidase B1/CPB1 Mouse anti-Human, Clone: 438806, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2897
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13459765
  • £249.49 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Carboxypeptidase B1/CPB1 Monoclonal antibody MAB2897 Specifically detects Carboxypeptidase B1/CPB1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Carboxypeptidase B1/CPB1
438806
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CPB1
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B1/CPB1 His16-Tyr417 Accession # P15086
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1360
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P15086
carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, DKFZp779K1333, EC 3.4.17, EC 3.4.17.2, Pancreas-specific protein, PASP, PCPB, procarboxypeptidase B
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Carboxypeptidase B1/CPB1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA2, rhCPE, recombinant mouse (rm) CPA4, and rmCPB1 is observed.