Learn More
Carboxypeptidase A2/CPA2 Mouse anti-Human, Clone: 384023, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB28961
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13449765
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Carboxypeptidase A2/CPA2 Monoclonal antibody MAB28961 Specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
Carboxypeptidase A2/CPA2
384023
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CPA2
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A2/CPA2 Leu18-His417 Accession # P48052
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1358
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P48052
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Carboxypeptidase A2/CPA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA4, rhCPB1, or rhCPE is observed. Recognizes the pro form (aa 17-417), but not the activated enzyme (aa 113-417) in Western blots.