missing translation for 'onlineSavingsMsg'
Learn More

Cathelicidin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen OSC00039W

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11580463

  • £257.00 / 150┬ÁL


Estimated Shipment
14-06-2018
Add to basket

Description

Description

OSC00039W is a control antibody for products OSC00037W, OSC00038G, OSC00009W, and OSC00028G.
Specifications

Specifications

CAP-18
Polyclonal
whole serum with no preservative
P49913
CAMP, CAP18, FALL39, FALL-39, 18kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Rabbit
150μL
Antibody
Polyclonal
Human
Control
Unconjugated
CAMP
Serum
CAMP
A synthetic scrambled peptide form of human Cathelicidin antimicrobial peptide (CAP-18, hCAP-18, antibacterial protein LL-37, CAMP, CRAMP, FALL39) conjugated to an immunogenic carrier protein has been used as the antigen
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
820
Videos