missing translation for 'onlineSavingsMsg'
Learn More

Cathelicidin, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen OSC00038G

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11570463

  • £257.00 / 500µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

Reconstitute with 500 ul of distilled water.
Specifications

Specifications

CAP-18
Polyclonal
Unconjugated
CAMP
Purified
CAMP
A synthetic peptide from human Cathelicidin antimicrobial peptide
Ammonium sulfate precipitation
avoiding freeze/thaw cycles., store at -20°C, store at 4°C short term. For long term storage
Primary
820
Western Blot
1mg/mL
Antibody with no preservative
P49913
CAMP, CAP18, FALL39, FALL-39, 18kDa cationic antimicrobial protein, CAP-18, hCAP-18, LL37, CRAMP, HSD26, hCAP18
Sheep
IgG
500μg
Antibody
Polyclonal
Human
Videos