Learn More
CNR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1746A
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-746A detects cannabinoid receptor 2 (CB2) from human and rat tissues as well as transfected rat CB2. PA1-746A has been successfully used in WB, immunocytochemistry, immunohistochemistry (paraffin) and FACS procedures. Immunocytochemical staining of CB2 into AtT20 cells transfected with the rat CB2 gene with PA1-746 yields a pattern consistent with plasma membrane staining.
CNR2 Polyclonal antibody specifically detects CNR2 in Human, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
CNR2 | |
Polyclonal | |
Unconjugated | |
CNR2 | |
Cannabinoid Receptor 2; cannabinoid receptor 2 (macrophage); CB2; CB-2; CB2 receptor; CB2A; CB2B; CB2C; CB2R; CB2-R; CNR2; CNR2C; CNR3; CX5; HCB2; HGNC:2160; OTTHUMP00000044841; peripheral cannabinoid receptor; rCB2; RP11-4M23.1; testis-dominant CNR2 isoform CB2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
1269, 57302 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
P34972, Q9QZN9 | |
CNR2 | |
GST fusion protein containing the first 32 amino acid residues from Rat CB2. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |