missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNR2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518428
Code : LD
Additional Details : Weight : 0.01000kg
Description
CNR2 Polyclonal antibody specifically detects CNR2 in Human samples. It is validated for Western Blot.Specifications
CNR2 | |
Polyclonal | |
Unconjugated | |
CNR2 | |
Cannabinoid Receptor 2; cannabinoid receptor 2 (macrophage); CB2; CB-2; CB2 receptor; CB2A; CB2B; CB2C; CB2R; CB2-R; CNR2; CNR2C; CNR3; CX5; HCB2; HGNC:2160; OTTHUMP00000044841; peripheral cannabinoid receptor; rCB2; RP11-4M23.1; testis-dominant CNR2 isoform CB2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
1269 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P34972 | |
CNR2 | |
Synthetic peptide sequence (TETEADGKITPWPD) corresponding to the C-terminus amino acids of CNR2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |