missing translation for 'onlineSavingsMsg'
Learn More

/ Parvovirus Mouse anti-Virus, Clone: CPV1-2A1, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA191114

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 11521112

  • £443.00 / 250µg

Estimated Shipment date
from Supplier 24-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CPV is reported to be a host range variant of FPV which emerged as a result of mutations of the viral capsid.
Specifications

Specifications

Canine/Feline Parvovirus
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
CPV; parvo; FPV
Proprietary immunogen
Protein A
RUO
Antibody
Monoclonal
ELISA, Immunohistochemistry (Paraffin)
CPV1-2A1
Unconjugated
Liquid
Mouse
IgG2a
250 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
Virus
Videos
Documents