Learn More
Goat anti-Canine IgM Secondary Antibody, HRP, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen PA184638
Code : LD
Additional Details : Weight : 1.24740kg
Description
Goat anti Dog IgM polyclonal antibody recognizes canine IgM and shows no cross - reactivity with other canine immunoglobulin classes in immunoelectrophoresis and ELISA. Goat anti Dog IgM may cross react with IgM from other species.
Canine IgM Polyclonal antibody specifically detects Canine IgM in Canine samples. It is validated for ELISA.Specifications
Canine IgM | |
Polyclonal | |
HRP | |
Goat | |
Affinity chromatography | |
RUO | |
Canine | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.2% BSA and 0.05% ProClin 300 | |
Purified canine IgM. | |
1 mg | |
Secondary | |
4° C, do not freeze | |
IgG |